2-(3-Methyl-5-isoxazolyl)piperidine

Supplier: Matrix Scientific

053798-500MG 053798-2.5G
101905-048EA 364.62 USD
101905-048 101905-050
2-(3-Methyl-5-isoxazolyl)piperidine
2-(3-Methyl-5-isoxazolyl)piperidine

Hazard: Irritant

Formula: C₉H₁₄N₂O
MDL Number: MFCD13562915
CAS Number: 1211509-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR