2-Chloro-6-methoxybenzoic acid
Supplier: Matrix Scientific
053796-500MG
053796-2.5G
101905-040EA
364.62
USD
101905-040
101905-042
2-Chloro-6-methoxybenzoic acid
2-Chloro-6-methoxybenzoic acid
Hazard: Irritant
Formula:
C₈H₇ClO₃ |
MDL Number:
MFCD00127702 CAS Number: 3260-89-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...