Methyl-2-chloro-3-quinolinecarboxylate

Supplier: Matrix Scientific

053793-500MG 053793-2.5G
101905-032EA 364.62 USD
101905-032 101905-034
Methyl-2-chloro-3-quinolinecarboxylate
Methyl-2-chloro-3-quinolinecarboxylate

Hazard: Irritant

Formula: C₁₁H₈ClNO₂
MDL Number: MFCD06203870
CAS Number: 16498-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR