2-[(3-Pyridinyloxy)methyl]benzoic acid
Supplier: Matrix Scientific
053790-500MG
053790-2.5G
101905-020EA
364.62
USD
101905-020
101905-022
2-[(3-Pyridinyloxy)methyl]benzoic acid
2-[(3-Pyridinyloxy)methyl]benzoic acid
Hazard: Irritant
Formula:
C₁₃H₁₁NO₃ MW: 229.24 g/mol |
MDL Number:
MFCD13562912 CAS Number: 1220035-84-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...