2-[(2-Fluorophenoxy)methyl]benzoic acid
Supplier: Matrix Scientific
053789-500MG
053789-2.5G
101905-016EA
364.62
USD
101905-016
101905-018
2-[(2-Fluorophenoxy)methyl]benzoic acid
2-[(2-Fluorophenoxy)methyl]benzoic acid
Hazard: Irritant
Formula:
C₁₄H₁₁FO₃ |
MDL Number:
MFCD12072100 CAS Number: 1156944-71-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...