2-[(3-Fluorophenoxy)methyl]benzoic acid
Supplier: Matrix Scientific
053788-500MG
053788-2.5G
101905-012EA
364.62
USD
101905-012
101905-014
2-[(3-Fluorophenoxy)methyl]benzoic acid
2-[(3-Fluorophenoxy)methyl]benzoic acid
Hazard: Irritant
Formula:
C₁₄H₁₁FO₃ |
MDL Number:
MFCD12072102 CAS Number: 114312-47-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...