2-[(4-Methylphenoxy)methyl]benzoic acid

Supplier: Matrix Scientific

053787-500MG 053787-2.5G
101905-008EA 364.62 USD
101905-008 101905-010
2-[(4-Methylphenoxy)methyl]benzoic acid
2-[(4-Methylphenoxy)methyl]benzoic acid

Hazard: Irritant

Formula: C₁₅H₁₄O₃
MDL Number: MFCD00963955
CAS Number: 728-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR