2-[(2-Methylphenoxy)methyl]benzoic acid
Supplier: Matrix Scientific
053786-500MG
053786-2.5G
101905-004EA
364.62
USD
101905-004
101905-006
2-[(2-Methylphenoxy)methyl]benzoic acid
2-[(2-Methylphenoxy)methyl]benzoic acid
Hazard: Irritant
Formula:
C₁₅H₁₄O₃ |
MDL Number:
MFCD05662414 CAS Number: 108475-90-7 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...