4-[(2-Fluorophenoxy)methyl]benzoic acid
Supplier: Matrix Scientific
053784-500MG
053784-2.5G
101904-996EA
364.62
USD
101904-996
101904-998
4-[(2-Fluorophenoxy)methyl]benzoic acid
4-[(2-Fluorophenoxy)methyl]benzoic acid
Hazard: Irritant
Formula:
C₁₄H₁₁FO₃ |
MDL Number:
MFCD09741132 CAS Number: 943114-74-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...