3-[(3,5-Dimethylphenoxy)methyl]benzoic acid
Supplier: Matrix Scientific
053783-500MG
053783-2.5G
101904-992EA
461.54
USD
101904-992
101904-994
3-[(3,5-Dimethylphenoxy)methyl]benzoic acid
3-[(3,5-Dimethylphenoxy)methyl]benzoic acid
Hazard: Irritant
Formula:
C₁₆H₁₆O₃ |
MDL Number:
MFCD09706921 CAS Number: 938140-74-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...