3-[(3-Fluorophenoxy)methyl]benzoic acid
Supplier: Matrix Scientific
053781-500MG
053781-2.5G
101904-984EA
364.62
USD
101904-984
101904-986
3-[(3-Fluorophenoxy)methyl]benzoic acid
3-[(3-Fluorophenoxy)methyl]benzoic acid
Hazard: Irritant
Formula:
C₁₄H₁₁FO₃ |
MDL Number:
MFCD11522685 CAS Number: 1019462-57-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...