3-[(2-Fluorophenoxy)methyl]benzoic acid
Supplier: Matrix Scientific
053780-500MG
053780-2.5G
101904-980EA
461.54
USD
101904-980
101904-982
3-[(2-Fluorophenoxy)methyl]benzoic acid
3-[(2-Fluorophenoxy)methyl]benzoic acid
Hazard: Irritant
Formula:
C₁₄H₁₁FO₃ |
MDL Number:
MFCD09755755 CAS Number: 944891-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...