3-(Phenoxymethyl)benzoic acid

Supplier: Matrix Scientific

053779-500MG 053779-2.5G
101904-976EA 364.62 USD
101904-976 101904-978
3-(Phenoxymethyl)benzoic acid
3-(Phenoxymethyl)benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₂O₃
MDL Number: MFCD00436828
CAS Number: 31719-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR