3-(Ethoxymethyl)benzoic acid
Supplier: Matrix Scientific
053777-500MG
053777-2.5G
101904-968EA
461.54
USD
101904-968
101904-970
3-(Ethoxymethyl)benzoic acid
3-(Ethoxymethyl)benzoic acid
Hazard: Irritant
Formula:
C₁₀H₁₂O₃ |
MDL Number:
MFCD09948208 CAS Number: 174894-85-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...