3-(Pyrrolidin-3-yl)pyridine dihydrochloride

Supplier: Matrix Scientific

Synonyms: 3-(3-Pyrrolidinyl)pyridine dihydrochloride

053771-500MG 053771-2.5G
101904-950EA 592.31 USD
101904-950 101904-952
3-(Pyrrolidin-3-yl)pyridine dihydrochloride
3-(Pyrrolidin-3-yl)pyridine dihydrochloride

Hazard: Irritant

Formula: C₉H₁₄Cl₂N₂
MW: 221.127 g/mol
MDL Number: MFCD02089474
CAS Number: 635309-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR