Methyl-2-(3-piperidinyl)benzoate hydrochloride
Supplier: Matrix Scientific
053769-500MG
053769-2.5G
101904-942EA
364.62
USD
101904-942
101904-944
Methyl-2-(3-piperidinyl)benzoate hydrochloride
Methyl-2-(3-piperidinyl)benzoate hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₈ClNO₂ MW: 255.75 g/mol |
MDL Number:
MFCD08751527 CAS Number: 1203682-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...