2-Pyrid-3-ylbenzoic acid
Supplier: Matrix Scientific
Synonyms:
2-(Pyridin-3-yl)benzoic acid
053768-500MG
053768-2.5G
101904-938EA
130.77
USD
101904-938
101904-940
2-Pyrid-3-ylbenzoic acid
2-Pyrid-3-ylbenzoic acid
Hazard: Irritant
Formula:
C₁₂H₉NO₂ MW: 199.2054 g/mol |
MDL Number:
MFCD03426511 CAS Number: 134363-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...