1-Naphthyl(1-piperazinyl)methanone

Supplier: Matrix Scientific

053765-500MG 053765-2.5G
101904-926EA 364.62 USD
101904-926 101904-928
1-Naphthyl(1-piperazinyl)methanone
1-Naphthyl(1-piperazinyl)methanone

Hazard: Irritant

Formula: C₁₅H₁₆N₂O
MDL Number: MFCD02256021
CAS Number: 684249-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR