1-Piperazinyl(2-thienyl)methanone
Supplier: Matrix Scientific
053764-500MG
053764-2.5G
101904-922EA
364.62
USD
101904-922
101904-924
1-Piperazinyl(2-thienyl)methanone
1-Piperazinyl(2-thienyl)methanone
Hazard: Irritant
Formula:
C₉H₁₂N₂OS |
MDL Number:
MFCD01807000 CAS Number: 52063-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...