(3,5-Difluorophenyl)(1-piperazinyl)methanone

Supplier: Matrix Scientific

053763-500MG 053763-2.5G
101904-918EA 364.62 USD
101904-918 101904-920
(3,5-Difluorophenyl)(1-piperazinyl)methanone
(3,5-Difluorophenyl)(1-piperazinyl)methanone

Hazard: Irritant

Formula: C₁₁H₁₂F₂N₂O
MDL Number: MFCD08443571
CAS Number: 874942-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR