(3-Methylphenyl)(1-piperazinyl)methanone
Supplier: Matrix Scientific
Synonyms:
Piperazin-1-yl(m-tolyl)methanone
053761-500MG
053761-2.5G
101904-910EA
364.62
USD
101904-910
101904-912
(3-Methylphenyl)(1-piperazinyl)methanone
(3-Methylphenyl)(1-piperazinyl)methanone
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O MW: 204.27 g/mol |
MDL Number:
MFCD04116565 CAS Number: 100939-91-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...