(3-Methoxyphenyl)(1-piperazinyl)methanone
Supplier: Matrix Scientific
053760-500MG
053760-2.5G
101904-906EA
364.62
USD
101904-906
101904-908
(3-Methoxyphenyl)(1-piperazinyl)methanone
(3-Methoxyphenyl)(1-piperazinyl)methanone
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₂ |
MDL Number:
MFCD01829255 CAS Number: 100939-89-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...