(4-Fluorophenyl)(1-piperazinyl)methanone

Supplier: Matrix Scientific

053759-500MG 053759-2.5G
101904-902EA 270.77 USD
101904-902 101904-904
(4-Fluorophenyl)(1-piperazinyl)methanone
(4-Fluorophenyl)(1-piperazinyl)methanone

Hazard: Irritant

Formula: C₁₁H₁₃FN₂O
MDL Number: MFCD01829194
CAS Number: 102391-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR