1-Piperazinyl(4-pyridinyl)methanone hydrochloride

Supplier: Matrix Scientific

053757-500MG 053757-2.5G
101904-894EA 769.23 USD
101904-894 101904-896
1-Piperazinyl(4-pyridinyl)methanone hydrochloride
1-Piperazinyl(4-pyridinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃O
MDL Number: MFCD19442202
CAS Number: 39640-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR