1-Piperazinyl(2-pyridinyl)methanone hydrochloride
Supplier: Matrix Scientific
053755-500MG
053755-2.5G
101904-886EA
538.46
USD
101904-886
101904-888
1-Piperazinyl(2-pyridinyl)methanone hydrochloride
1-Piperazinyl(2-pyridinyl)methanone hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₄ClN₃O |
MDL Number:
MFCD09971513 CAS Number: 1071608-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...