tert-Butyl (1R,2R)-2-(benzylamino)cyclohexylcarbamate

Supplier: Matrix Scientific

053752-500MG 053752-2.5G
101904-874EA 364.62 USD
101904-874 101904-876
tert-Butyl (1R,2R)-2-(benzylamino)cyclohexylcarbamate
tert-Butyl (1R,2R)-2-(benzylamino)cyclohexylcarbamate

Hazard: Irritant

Formula: C₁₈H₂₈N₂O₂
MW: 304.44 g/mol
MDL Number: MFCD09054925
CAS Number: 1217764-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR