4-[(Tetrahydro-2-furanylmethyl)amino]-2-pyrimidinol
Supplier: Matrix Scientific
053750-500MG
053750-2.5G
101904-866EA
364.62
USD
101904-866
101904-868
4-[(Tetrahydro-2-furanylmethyl)amino]-2-pyrimidinol
4-[(Tetrahydro-2-furanylmethyl)amino]-2-pyrimidinol
Hazard: Irritant
Formula:
C₉H₁₃N₃O₂ |
MDL Number:
MFCD09054907 CAS Number: 927803-70-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...