4-(Methylamino)-2-pyrimidinol
Supplier: Matrix Scientific
053749-500MG
053749-2.5G
101904-862EA
364.62
USD
101904-862
101904-864
4-(Methylamino)-2-pyrimidinol
4-(Methylamino)-2-pyrimidinol
Hazard: Irritant
Formula:
C₅H₇N₃O Melting Pt: 270 °C |
MDL Number:
MFCD09054904 CAS Number: 6220-47-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...