2-(1-Pyrrolidinyl)-5-pyrimidinecarboxylic acid

Supplier: Matrix Scientific

Synonyms: 2-(pyrrolidin-1-yl)pyrimidine-5-carboxylic acid

053747-500MG 053747-2.5G
101904-854EA 364.62 USD
101904-854 101904-856
2-(1-Pyrrolidinyl)-5-pyrimidinecarboxylic acid
2-(1-Pyrrolidinyl)-5-pyrimidinecarboxylic acid

Hazard: Irritant

Formula: C₉H₁₁N₃O₂
MW: 193.21 g/mol
MDL Number: MFCD09054869
CAS Number: 253315-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR