2-(Diethylamino)-5-pyrimidinecarboxylic acid
Supplier: Matrix Scientific
053746-500MG
053746-2.5G
101904-850EA
364.62
USD
101904-850
101904-852
2-(Diethylamino)-5-pyrimidinecarboxylic acid
2-(Diethylamino)-5-pyrimidinecarboxylic acid
Hazard: Irritant
Formula:
C₉H₁₃N₃O₂ |
MDL Number:
MFCD09054868 CAS Number: 927803-51-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...