Methyl-2-(benzylamino)-5-pyrimidinecarboxylate

Supplier: Matrix Scientific

053745-500MG 053745-2.5G
101904-846EA 364.62 USD
101904-846 101904-848
Methyl-2-(benzylamino)-5-pyrimidinecarboxylate
Methyl-2-(benzylamino)-5-pyrimidinecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₃N₃O₂
MDL Number: MFCD09054865
CAS Number: 927803-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR