(5-Chloro-2-methoxyphenyl)methanamine
Supplier: Matrix Scientific
053742-500MG
053742-2.5G
101904-834EA
247.69
USD
101904-834
101904-836
(5-Chloro-2-methoxyphenyl)methanamine
(5-Chloro-2-methoxyphenyl)methanamine
Hazard: Irritant
Formula:
C₈H₁₀ClNO |
MDL Number:
MFCD03410973 CAS Number: 181473-92-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...