3-Chloro-4-methoxyphenethylamine
Supplier: Matrix Scientific
053741-500MG
053741-2.5G
101904-830EA
364.62
USD
101904-830
101904-832
3-Chloro-4-methoxyphenethylamine
3-Chloro-4-methoxyphenethylamine
Hazard: Irritant
Formula:
C₉H₁₂ClNO |
MDL Number:
MFCD00996131 CAS Number: 7569-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...