(5-Chloro-2-thienyl)methanamine hydrochloride

Supplier: Matrix Scientific

053739-500MG 053739-2.5G
101904-822EA 364.62 USD
101904-822 101904-824
(5-Chloro-2-thienyl)methanamine hydrochloride
(5-Chloro-2-thienyl)methanamine hydrochloride

Hazard: Irritant

Formula: C₅H₇Cl₂NS
MDL Number: MFCD09971558
CAS Number: 548772-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR