3-(2-Methoxyphenoxy)propionic acid

Supplier: Matrix Scientific

053736-500MG 053736-2.5G
101904-810EA 364.62 USD
101904-810 101904-812
3-(2-Methoxyphenoxy)propionic acid
3-(2-Methoxyphenoxy)propionic acid

Hazard: Irritant

Formula: C₁₀H₁₂O₄
MW: 196.1999 g/mol
MDL Number: MFCD01931914
CAS Number: 20370-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR