3-(4-Methoxyphenoxy)propionic acid
Supplier: Matrix Scientific
053735-500MG
053735-2.5G
101904-806EA
364.62
USD
101904-806
101904-808
3-(4-Methoxyphenoxy)propionic acid
3-(4-Methoxyphenoxy)propionic acid
Hazard: Irritant
Formula:
C₁₀H₁₂O₄ |
MDL Number:
MFCD00020515 CAS Number: 20811-60-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...