3-(3-Methylphenoxy)propionic acid

Supplier: Matrix Scientific

053734-500MG 053734-2.5G
101904-802EA 76.92 USD
101904-802 101904-804
3-(3-Methylphenoxy)propionic acid
3-(3-Methylphenoxy)propionic acid

Hazard: Irritant

Formula: C₁₀H₁₂O₃
MDL Number: MFCD00642411
CAS Number: 25173-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR