Methyl-5-amino-2-[3-(trifluoromethyl)phenoxy]benzoate

Supplier: Matrix Scientific

053730-500MG 053730-2.5G
101904-790EA 364.62 USD
101904-790 101904-792
Methyl-5-amino-2-[3-(trifluoromethyl)phenoxy]benzoate
Methyl-5-amino-2-[3-(trifluoromethyl)phenoxy]benzoate

Hazard: Irritant

Formula: C₁₅H₁₂F₃NO₃
MDL Number: MFCD09054797
CAS Number: 927802-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR