Methyl-3-amino-4-phenoxybenzoate

Supplier: Matrix Scientific

053728-500MG 053728-2.5G
101904-782EA 364.62 USD
101904-782 101904-784
Methyl-3-amino-4-phenoxybenzoate
Methyl-3-amino-4-phenoxybenzoate

Hazard: Irritant

Formula: C₁₄H₁₃NO₃
MDL Number: MFCD09054789
CAS Number: 227275-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR