Methyl-3-amino-4-(cyclopropylmethoxy)benzoate

Supplier: Matrix Scientific

053727-500MG 053727-2.5G
101904-778EA 364.62 USD
101904-778 101904-780
Methyl-3-amino-4-(cyclopropylmethoxy)benzoate
Methyl-3-amino-4-(cyclopropylmethoxy)benzoate

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MDL Number: MFCD09054788
CAS Number: 927802-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR