Methyl-3-amino-4-(benzyloxy)benzoate

Supplier: Matrix Scientific

053725-500MG 053725-2.5G
101904-770EA 364.62 USD
101904-770 101904-772
Methyl-3-amino-4-(benzyloxy)benzoate
Methyl-3-amino-4-(benzyloxy)benzoate

Hazard: Irritant

Formula: C₁₅H₁₅NO₃
MDL Number: MFCD09054786
CAS Number: 927802-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR