Methyl-3-amino-4-isopropoxybenzoate

Supplier: Matrix Scientific

053724-500MG 053724-2.5G
101904-766EA 364.62 USD
101904-766 101904-768
Methyl-3-amino-4-isopropoxybenzoate
Methyl-3-amino-4-isopropoxybenzoate

Hazard: Irritant

Formula: C₁₁H₁₅NO₃
MDL Number: MFCD09054785
CAS Number: 927802-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR