Methyl-3-amino-4-ethoxybenzoate
Supplier: Matrix Scientific
053723-500MG
053723-2.5G
101904-762EA
461.54
USD
101904-762
101904-764
Methyl-3-amino-4-ethoxybenzoate
Methyl-3-amino-4-ethoxybenzoate
Hazard: Irritant
Formula:
C₁₀H₁₃NO₃ |
MDL Number:
MFCD09054784 CAS Number: 927802-54-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...