4-(Cyclohexyloxy)-1H-indazol-3-amine
Supplier: Matrix Scientific
053721-500MG
053721-2.5G
101904-754EA
364.62
USD
101904-754
101904-756
4-(Cyclohexyloxy)-1H-indazol-3-amine
4-(Cyclohexyloxy)-1H-indazol-3-amine
Hazard: Irritant
Formula:
C₁₃H₁₇N₃O |
MDL Number:
MFCD09054763 CAS Number: 927802-30-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...