4-(1H-Pyrazol-1-yl)-1H-indazol-3-ylamine
Supplier: Matrix Scientific
053720-500MG
053720-2.5G
101904-750EA
461.54
USD
101904-750
101904-752
4-(1H-Pyrazol-1-yl)-1H-indazol-3-ylamine
4-(1H-Pyrazol-1-yl)-1H-indazol-3-ylamine
Hazard: Irritant
Formula:
C₁₀H₉N₅ |
MDL Number:
MFCD09054758 CAS Number: 1214622-52-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...