1-Ethyl-1H-indazol-3-amine

Supplier: Matrix Scientific

053719-500MG 053719-2.5G
101904-746EA 364.62 USD
101904-746 101904-748
1-Ethyl-1H-indazol-3-amine
1-Ethyl-1H-indazol-3-amine

Hazard: Irritant

Formula: C₉H₁₁N₃
MDL Number: MFCD09054756
CAS Number: 927802-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR