3,4-Dimethyl-2-(1H-pyrazol-1-yl)benzoic acid

Supplier: Matrix Scientific

053717-500MG 053717-2.5G
101904-738EA 461.54 USD
101904-738 101904-740
3,4-Dimethyl-2-(1H-pyrazol-1-yl)benzoic acid
3,4-Dimethyl-2-(1H-pyrazol-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂
MDL Number: MFCD09054735
CAS Number: 1214622-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR