3,5-Dimethyl-2-(1H-pyrazol-1-yl)benzoic acid
Supplier: Matrix Scientific
053714-500MG
053714-2.5G
101904-726EA
364.62
USD
101904-726
101904-728
3,5-Dimethyl-2-(1H-pyrazol-1-yl)benzoic acid
Hazard: Irritant
Formula:
C₁₂H₁₂N₂O₂ |
MDL Number:
MFCD09054730 CAS Number: 1214622-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...