5-Fluoro-2-(1H-pyrazol-1-yl)benzoic acid

Supplier: Matrix Scientific

053712-500MG 053712-2.5G
101904-718EA 461.54 USD
101904-718 101904-720
5-Fluoro-2-(1H-pyrazol-1-yl)benzoic acid
5-Fluoro-2-(1H-pyrazol-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₀H₇FN₂O₂
MDL Number: MFCD09054728
CAS Number: 1152964-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR