3-Ethyl-2-(1H-pyrazol-1-yl)benzaldehyde

Supplier: Matrix Scientific

053711-500MG 053711-2.5G
101904-714EA 364.62 USD
101904-714 101904-716
3-Ethyl-2-(1H-pyrazol-1-yl)benzaldehyde
3-Ethyl-2-(1H-pyrazol-1-yl)benzaldehyde

Hazard: Irritant

Formula: C₁₂H₁₂N₂O
MDL Number: MFCD09054727
CAS Number: 1214622-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR